Product Details

SNP ID
rs34361429
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:62808604 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTCTCTATCAAAACTACTGAACT[C/T]CTGCACGACCTCATGAAAGCCACAT
Phenotype
MIM: 120270 MIM: 611157 MIM: 606459
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COL9A3 PubMed Links

Gene Details

Gene
COL9A3
Gene Name
collagen type IX alpha 3
There are no transcripts associated with this gene.

Gene
MRGBP
Gene Name
MRG/MORF4L binding protein
There are no transcripts associated with this gene.

Gene
OGFR
Gene Name
opioid growth factor receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007346.2 Intron NP_031372.2
Gene
OGFR-AS1
Gene Name
OGFR antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details