Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001853.3 | 2360 | UTR 3 | NP_001844.3 | ||
XM_011528545.1 | 2360 | Intron | XP_011526847.1 | ||
XM_017027666.1 | 2360 | Intron | XP_016883155.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001301726.1 | 2360 | Intron | NP_001288655.1 | ||
NM_006602.3 | 2360 | UTR 3 | NP_006593.2 | ||
XM_005260185.3 | 2360 | Intron | XP_005260242.1 | ||
XM_011528497.2 | 2360 | Intron | XP_011526799.1 | ||
XM_011528498.2 | 2360 | Intron | XP_011526800.1 | ||
XM_011528499.1 | 2360 | Intron | XP_011526801.1 |