Product Details

SNP ID
rs910397
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.20:33707735 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TTGGAGGGACGGCTCTTGTTATAGA[C/T]GAGGAAGTCTGAGATGTCGTGCCAT
Phenotype
MIM: 616397
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PXMP4 PubMed Links

Gene Details

Gene
PXMP4
Gene Name
peroxisomal membrane protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007238.4 534 Missense Mutation ATC,GTC I204V NP_009169.3
NM_183397.2 534 UTR 3 NP_899634.1

View Full Product Details