Product Details

SNP ID
rs332233
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:64075546 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAATGTAGATTTCCAACTGGAAAG[C/T]GGGAAAAAAAGTAAAAGAAAAAACA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM96A PubMed Links

Gene Details

Gene
FAM96A
Gene Name
family with sequence similarity 96 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014812.2 Intron NP_001014812.1
NM_001289108.1 Intron NP_001276037.1
NM_032231.5 Intron NP_115607.1

View Full Product Details