Product Details

SNP ID
rs3739144
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:79085288 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATCCTTCCATTTCTTGAATCCTAT[A/G]GTACAATGAGAAGTGTCAGACATAG
Phenotype
MIM: 167771
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
REG1B PubMed Links

Gene Details

Gene
REG1B
Gene Name
regenerating family member 1 beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006507.3 Intron NP_006498.1

View Full Product Details