Product Details
- SNP ID
-
rs35943476
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
18
- Location
-
Chr.1:17313873 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGATAACACATGTGGGTCACACGC[C/T]TGAGGGCATGCACACACCTGGTGTG
- Phenotype
-
MIM: 605347
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PADI4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78966846] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PADI4
- Gene Name
- peptidyl arginine deiminase 4
View Full Product Details