Product Details

SNP ID
rs6817138
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.4:139303347 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATAGCAATTTATCTTTAATCTCTT[C/T]TTCACAATTCTTTATTTTGAAAAAA
Phenotype
MIM: 608000 MIM: 603844
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
NAA15 PubMed Links

Gene Details

Gene
NAA15
Gene Name
N(alpha)-acetyltransferase 15, NatA auxiliary subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_057175.3 Intron NP_476516.1
XM_005263236.2 Intron XP_005263293.1
Gene
NDUFC1
Gene Name
NADH:ubiquinone oxidoreductase subunit C1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184986.1 Intron NP_001171915.1
NM_001184987.1 Intron NP_001171916.1
NM_001184988.1 Intron NP_001171917.1
NM_001184989.1 Intron NP_001171918.1
NM_001184990.1 Intron NP_001171919.1
NM_001184991.1 Intron NP_001171920.1
NM_002494.3 Intron NP_002485.1

View Full Product Details