Product Details

SNP ID
rs4936845
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:123805523 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGGTTCAGGAGAGGGGTCACCACT[G/T]TGATGAGGACAGCGGCCACCTTGTC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR6M1 PubMed Links

Gene Details

Gene
OR6M1
Gene Name
olfactory receptor family 6 subfamily M member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005325.1 827 Missense Mutation AAA,ACA K276T NP_001005325.1

View Full Product Details