Product Details
- SNP ID
-
rs10817199
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:111631995 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTGACATTGTTATGCAAAACTTTGG[C/G]CATCTTACGATTTCGTGAAAATATT
- Phenotype
-
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
DNAJC25
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75856829] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DNAJC25
- Gene Name
- DnaJ heat shock protein family (Hsp40) member C25
- Gene
- DNAJC25-GNG10
- Gene Name
- DNAJC25-GNG10 readthrough
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004125.3 |
|
Intron |
|
|
NP_004116.2 |
View Full Product Details