Product Details

SNP ID
rs2234528
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:100337318 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTGAGTGCTGTCAGATTCTGGACC[A/C]GGGAAAGGCCAAGGCGCACAGCCGG
Phenotype
MIM: 609911 MIM: 191050
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SLC25A47 PubMed Links

Gene Details

Gene
SLC25A47
Gene Name
solute carrier family 25 member 47
There are no transcripts associated with this gene.

Gene
WARS
Gene Name
tryptophanyl-tRNA synthetase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004184.3 Intron NP_004175.2
NM_173701.1 Intron NP_776049.1
NM_213645.1 Intron NP_998810.1
NM_213646.1 Intron NP_998811.1
XM_005268044.3 Intron XP_005268101.1
XM_006720249.3 Intron XP_006720312.1
XM_011537133.2 Intron XP_011535435.1
XM_011537135.2 Intron XP_011535437.1
XM_011537136.2 Intron XP_011535438.1
XM_017021627.1 Intron XP_016877116.1
XM_017021628.1 Intron XP_016877117.1
XM_017021629.1 Intron XP_016877118.1

View Full Product Details