Product Details

SNP ID
rs1940621
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.18:5954988 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
AATTCTCCAAGCAGAATCAACTTTC[C/T]TAAACCTTAACATTCTTAATGAAGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
L3MBTL4 PubMed Links

Gene Details

Gene
L3MBTL4
Gene Name
l(3)mbt-like 4 (Drosophila)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173464.3 3399 UTR 3 NP_775735.2
XM_005258166.1 3399 UTR 3 XP_005258223.1
XM_006722364.2 3399 Intron XP_006722427.1
XM_011525757.1 3399 UTR 3 XP_011524059.1
XM_011525758.1 3399 UTR 3 XP_011524060.1
XM_011525759.1 3399 UTR 3 XP_011524061.1
XM_011525760.1 3399 UTR 3 XP_011524062.1
XM_011525761.1 3399 UTR 3 XP_011524063.1
XM_011525762.1 3399 Intron XP_011524064.1
XM_011525763.2 3399 Intron XP_011524065.1
XM_011525765.2 3399 Intron XP_011524067.1
XM_011525767.2 3399 UTR 3 XP_011524069.1
XM_017026074.1 3399 UTR 3 XP_016881563.1
XM_017026075.1 3399 UTR 3 XP_016881564.1
XM_017026076.1 3399 UTR 3 XP_016881565.1
XM_017026077.1 3399 Intron XP_016881566.1

View Full Product Details