Product Details

SNP ID
rs2167689
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.18:33854522 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TTTGCAAACAGAAGTTACTGAACCT[A/G]TGTTGAGGCAAATCATTAACATCAT
Phenotype
MIM: 603577
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NOL4 PubMed Links

Gene Details

Gene
NOL4
Gene Name
nucleolar protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198546.1 Intron NP_001185475.1
NM_001198547.1 Intron NP_001185476.1
NM_001198548.1 Intron NP_001185477.1
NM_001198549.1 Intron NP_001185478.1
NM_001282527.1 Intron NP_001269456.1
NM_003787.4 Intron NP_003778.2
XM_006722563.2 Intron XP_006722626.1
XM_006722564.2 Intron XP_006722627.1
XM_006722565.1 Intron XP_006722628.1
XM_011526237.1 Intron XP_011524539.1
XM_011526238.1 Intron XP_011524540.1
XM_011526239.2 Intron XP_011524541.1
XM_017026052.1 Intron XP_016881541.1
XM_017026053.1 Intron XP_016881542.1
XM_017026054.1 Intron XP_016881543.1
XM_017026055.1 Intron XP_016881544.1
XM_017026056.1 Intron XP_016881545.1
XM_017026057.1 Intron XP_016881546.1
XM_017026058.1 Intron XP_016881547.1
XM_017026059.1 Intron XP_016881548.1
XM_017026060.1 Intron XP_016881549.1

View Full Product Details