Product Details
- SNP ID
-
rs2282623
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:57234352 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AATTTGAAGCCAGGCGGGGAGGCAC[C/T]GGAAAGGGGCCAAGGGAGGCTCTCA
- Phenotype
-
MIM: 600052
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
APLNR
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79024304] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- APLNR
- Gene Name
- apelin receptor
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005161.4 |
3102 |
UTR 3 |
|
|
NP_005152.1 |
View Full Product Details