Product Details
- SNP ID
-
rs2282624
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:57234437 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGAGCAGGAACCCAGCTCAGTAGGA[C/T]GCCCCCAAGCCTTGGCGCCAGCTGC
- Phenotype
-
MIM: 600052
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
APLNR
PubMed Links
Gene Details
- Gene
- APLNR
- Gene Name
- apelin receptor
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005161.4 |
3017 |
UTR 3 |
|
|
NP_005152.1 |
View Full Product Details