Product Details

SNP ID
rs1985242
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:113977551 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGCCAGATCTCTGCTTTCCTGTCA[A/T]TGAATGCATAGGTCCTTTCTACAAG
Phenotype
MIM: 182139
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
HTR3A PubMed Links

Gene Details

Gene
HTR3A
Gene Name
5-hydroxytryptamine receptor 3A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000869.5 52 Intron NP_000860.2
NM_001161772.2 52 UTR 5 NP_001155244.1
NM_213621.3 52 Intron NP_998786.2

View Full Product Details