Product Details
- SNP ID
-
rs11635081
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:85761029 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCGCGGTCTGTGGAGGGAGGAGAG[A/G]AAGAGAAAATCAATCAGTGGCCACA
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
KLHL25
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs117972215] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KLHL25
- Gene Name
- kelch like family member 25
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_022480.3 |
|
Intron |
|
|
NP_071925.2 |
- Gene
- LOC101929679
- Gene Name
- uncharacterized LOC101929679
There are no transcripts associated with this gene.
- Gene
- MIR1276
- Gene Name
- microRNA 1276
There are no transcripts associated with this gene.
View Full Product Details