Product Details

SNP ID
rs3743336
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:85759504 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGGCCAGGCTCCTGAGTGTGCCAGC[A/T]GAGCCGTCCCCTGGGACCACAGCCA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
KLHL25 PubMed Links

Gene Details

Gene
KLHL25
Gene Name
kelch like family member 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022480.3 3505 UTR 3 NP_071925.2
Gene
LOC101929679
Gene Name
uncharacterized LOC101929679
There are no transcripts associated with this gene.

View Full Product Details