Product Details

SNP ID
rs2161612
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:39106998 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAATATAAGATATAACCATAAGTAG[A/G]AGTATAAACAATAATTTTTCTTCTT
Phenotype
MIM: 602731
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FYB PubMed Links

Gene Details

Gene
FYB
Gene Name
FYN binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243093.1 2921 UTR 3 NP_001230022.1
NM_001465.4 2921 UTR 3 NP_001456.3
NM_199335.3 2921 UTR 3 NP_955367.1
XM_006714464.2 2921 UTR 3 XP_006714527.1
XM_006714465.2 2921 UTR 3 XP_006714528.1
XM_006714466.2 2921 UTR 3 XP_006714529.1
XM_011514008.2 2921 UTR 3 XP_011512310.1
XM_011514009.1 2921 UTR 3 XP_011512311.1
XM_011514010.1 2921 UTR 3 XP_011512312.1
XM_011514011.2 2921 UTR 3 XP_011512313.1
XM_011514012.2 2921 Intron XP_011512314.1
XM_011514013.2 2921 Intron XP_011512315.1
XM_017009316.1 2921 UTR 3 XP_016864805.1

View Full Product Details