Product Details

SNP ID
rs12649314
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:98277029 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTTTAGTTAGGGGCAGTGTCAGGC[A/C]TAGAATGTTGATCTCTCAACTCCTA
Phenotype
MIM: 179502
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RAP1GDS1 PubMed Links

Gene Details

Gene
RAP1GDS1
Gene Name
Rap1 GTPase-GDP dissociation stimulator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100426.1 Intron NP_001093896.1
NM_001100427.1 Intron NP_001093897.1
NM_001100428.1 Intron NP_001093898.1
NM_001100429.1 Intron NP_001093899.1
NM_001100430.1 Intron NP_001093900.1
NM_021159.4 Intron NP_066982.3

View Full Product Details