Product Details

SNP ID
rs9782
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.12:102958048 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CAGGGAGGAGAAAAAGCATTTTCAC[C/T]TTTTTTGCTCCCACTCTAAGAAGTC
Phenotype
MIM: 100790
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ASCL1 PubMed Links

Gene Details

Gene
ASCL1
Gene Name
achaete-scute family bHLH transcription factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004316.3 375 UTR 5 NP_004307.2

View Full Product Details