Product Details

SNP ID
rs3363
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.20:45948675 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AAGTAAGCTGTTCAGGACTGGACTC[C/T]GGTCCCTTTATTGAGACTGACAGGC
Phenotype
MIM: 610827
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PCIF1 PubMed Links

Gene Details

Gene
PCIF1
Gene Name
PDX1 C-terminal inhibiting factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022104.3 4201 Intron NP_071387.1
XM_011528980.2 4201 Intron XP_011527282.1
XM_011528981.2 4201 Intron XP_011527283.1
XM_017028013.1 4201 Intron XP_016883502.1
XM_017028014.1 4201 Intron XP_016883503.1
Gene
ZNF335
Gene Name
zinc finger protein 335
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022095.3 4201 UTR 3 NP_071378.1
XM_005260504.4 4201 UTR 3 XP_005260561.1
XM_011528979.2 4201 Intron XP_011527281.1
XM_017028012.1 4201 UTR 3 XP_016883501.1

View Full Product Details