Product Details

SNP ID
rs883813
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:98691 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGTATGAATGTTTTGTATCTTTGT[A/G]TATGATCAGCTTAAAAGTATTCACT
Phenotype
MIM: 612118
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IQSEC3 PubMed Links

Gene Details

Gene
IQSEC3
Gene Name
IQ motif and Sec7 domain 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170738.1 Intron NP_001164209.1
NM_015232.1 Intron NP_056047.1
XM_011520958.2 Intron XP_011519260.1
XM_011520959.2 Intron XP_011519261.1
XM_011520960.1 Intron XP_011519262.1
XM_011520961.1 Intron XP_011519263.1
XM_017019310.1 Intron XP_016874799.1
XM_017019311.1 Intron XP_016874800.1

View Full Product Details