Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001284403.1 | Intron | NP_001271332.1 | |||
NM_001284404.1 | Intron | NP_001271333.1 | |||
NM_001284405.1 | Intron | NP_001271334.1 | |||
NM_024754.4 | Intron | NP_079030.3 | |||
XM_005248601.2 | Intron | XP_005248658.1 | |||
XM_005248603.2 | Intron | XP_005248660.1 | |||
XM_017009866.1 | Intron | XP_016865355.1 | |||
XM_017009867.1 | Intron | XP_016865356.1 | |||
XM_017009868.1 | Intron | XP_016865357.1 |