Product Details

SNP ID
rs3138500
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:89605030 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGGGCGCACTCGCTGGTGGTGGG[C/T]GCGCCGTGCTGAGCTCTGGCTGTCA
Phenotype
MIM: 604949
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GADD45G PubMed Links

Gene Details

Gene
GADD45G
Gene Name
growth arrest and DNA damage inducible gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006705.3 19 UTR 5 NP_006696.1
XM_011518163.2 19 UTR 5 XP_011516465.1

View Full Product Details