Product Details

SNP ID
rs13171053
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:172658593 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTGTGTTGACCTCCTTCCCTGCA[A/G]GGATGACTTCTGCACTTTGCCGTCA
Phenotype
MIM: 615893
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC100130394 PubMed Links

Gene Details

Gene
LOC100130394
Gene Name
translation machinery associated 7 homolog (S. cerevisiae) pseudogene
There are no transcripts associated with this gene.

Gene
MIR5003
Gene Name
microRNA 5003
There are no transcripts associated with this gene.

Gene
NEURL1B
Gene Name
neuralized E3 ubiquitin protein ligase 1B
There are no transcripts associated with this gene.

View Full Product Details