Product Details

SNP ID
rs2043305
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10617644 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCATGCTCCTGCTCAAACACCCTCC[A/G]TGGCTCCCACGTACTCATCAAACCT
Phenotype
MIM: 606106
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC44A2 PubMed Links

Gene Details

Gene
SLC44A2
Gene Name
solute carrier family 44 member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145056.1 Intron NP_001138528.1
NM_020428.3 Intron NP_065161.3
XM_005259997.1 Intron XP_005260054.1
XM_005259999.1 Intron XP_005260056.1

View Full Product Details