Product Details
- SNP ID
-
rs872478
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:16502561 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- GCATGGAAAAGTTTTTATCTCTTCT[C/G]TTTCCTCTCCTGTAAGTAAAGATTT
- Phenotype
-
MIM: 603450
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
LDB2
PubMed Links
Gene Details
- Gene
- LDB2
- Gene Name
- LIM domain binding 2
View Full Product Details