Product Details

SNP ID
rs34876489
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:27772714 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGCGCCATTTTTAATCCTGGCGA[C/G]GCGAAGTGTAGTGGCAAGGAGTTTG
Phenotype
MIM: 609353
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CCDC25 PubMed Links

Gene Details

Gene
CCDC25
Gene Name
coiled-coil domain containing 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304529.1 Intron NP_001291458.1
NM_001304530.1 Intron NP_001291459.1
NM_001304532.1 Intron NP_001291461.1
NM_018246.2 Intron NP_060716.2
XM_011544570.2 Intron XP_011542872.1
XM_011544571.2 Intron XP_011542873.1
Gene
ESCO2
Gene Name
establishment of sister chromatid cohesion N-acetyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017420.2 Intron NP_001017420.1
XM_011544421.2 Intron XP_011542723.1
XM_011544422.2 Intron XP_011542724.1

View Full Product Details