Product Details
- SNP ID
-
rs7672297
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:82633727 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTGGTTTTAATTTGCATTTCTCTAA[G/T]GATTAGTTCATTTGATTTTCATAAA
- Phenotype
-
MIM: 608370
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SCD5
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77902864] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SCD5
- Gene Name
- stearoyl-CoA desaturase 5
View Full Product Details