Product Details

SNP ID
rs360790
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:62688179 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTAATTTAATATTGGATGTAAAAA[C/T]TGGATATTTCCAATAGCATGAGAGA
Phenotype
MIM: 609922
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EHBP1 PubMed Links

Gene Details

Gene
EHBP1
Gene Name
EH domain binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142614.1 Intron NP_001136086.1
NM_001142615.2 Intron NP_001136087.1
NM_001142616.1 Intron NP_001136088.1
NM_015252.3 Intron NP_056067.2
XM_005264225.2 Intron XP_005264282.1
XM_005264226.2 Intron XP_005264283.1
XM_005264227.1 Intron XP_005264284.1
XM_005264228.2 Intron XP_005264285.1
XM_006711971.2 Intron XP_006712034.1
XM_011532713.2 Intron XP_011531015.1
XM_011532714.2 Intron XP_011531016.1
XM_011532715.2 Intron XP_011531017.1
XM_011532716.2 Intron XP_011531018.1
XM_011532717.1 Intron XP_011531019.1
XM_011532718.2 Intron XP_011531020.1
XM_017003643.1 Intron XP_016859132.1
XM_017003644.1 Intron XP_016859133.1
XM_017003645.1 Intron XP_016859134.1
XM_017003646.1 Intron XP_016859135.1
XM_017003647.1 Intron XP_016859136.1
XM_017003648.1 Intron XP_016859137.1
XM_017003649.1 Intron XP_016859138.1
XM_017003650.1 Intron XP_016859139.1
XM_017003651.1 Intron XP_016859140.1
XM_017003652.1 Intron XP_016859141.1
XM_017003653.1 Intron XP_016859142.1
XM_017003654.1 Intron XP_016859143.1
XM_017003655.1 Intron XP_016859144.1

View Full Product Details