Product Details

SNP ID
rs1044731
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:56958903 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACGAGAATAAGAAAGGTGGCTGAA[A/G]TAGATAATTTCAGTGACGGAGGGGA
Phenotype
MIM: 614010
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IMPAD1 PubMed Links

Gene Details

Gene
IMPAD1
Gene Name
inositol monophosphatase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017813.4 6256 UTR 3 NP_060283.3

View Full Product Details