Product Details

SNP ID
rs3853
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:22713331 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCATACCCACATCGTCGGTGGTG[A/G]GCAGTGTGGCCACATGCCCGGATGG
Phenotype
MIM: 612473
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PEBP4 PubMed Links

Gene Details

Gene
PEBP4
Gene Name
phosphatidylethanolamine binding protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144962.2 870 UTR 3 NP_659399.2
XM_011544414.2 870 UTR 3 XP_011542716.1
XM_017013103.1 870 Intron XP_016868592.1

View Full Product Details