Product Details

SNP ID
rs11610949
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:27334535 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGTCCATTAAGAAAGTGTTGTCCC[C/T]TGTGAGTTAAAATTGGCCAAACTGA
Phenotype
MIM: 614517 MIM: 615836
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARNTL2 PubMed Links

Gene Details

Gene
ARNTL2
Gene Name
aryl hydrocarbon receptor nuclear translocator like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001248002.1 Intron NP_001234931.1
NM_001248003.1 Intron NP_001234932.1
NM_001248004.1 Intron NP_001234933.1
NM_001248005.1 Intron NP_001234934.1
NM_020183.4 Intron NP_064568.3
XM_006719112.3 Intron XP_006719175.1
XM_006719114.3 Intron XP_006719177.1
XM_011520766.2 Intron XP_011519068.1
XM_011520768.2 Intron XP_011519070.1
XM_011520769.2 Intron XP_011519071.1
XM_011520770.2 Intron XP_011519072.1
XM_011520771.2 Intron XP_011519073.1
XM_017019666.1 Intron XP_016875155.1
XM_017019667.1 Intron XP_016875156.1
XM_017019668.1 Intron XP_016875157.1
XM_017019669.1 Intron XP_016875158.1
Gene
STK38L
Gene Name
serine/threonine kinase 38 like
There are no transcripts associated with this gene.

View Full Product Details