Product Details

SNP ID
rs10759610
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:97309128 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TACTCAGCCAATCCCTATTATTAGT[C/G]AGTTCTGTTTTTCCAATTTCTTGTG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CCDC180 PubMed Links

Gene Details

Gene
CCDC180
Gene Name
coiled-coil domain containing 180
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020893.2 Intron NP_065944.2
Gene
LOC100499484-C9ORF174
Gene Name
LOC100499484-C9orf174 readthrough
There are no transcripts associated with this gene.

View Full Product Details