Product Details

SNP ID
rs35852964
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:113357049 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGTGGGAGAGGATGACATCACTA[A/G]GTGAATGAGTGTAGACACGAAAGAG
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
BSPRY PubMed Links

Gene Details

Gene
BSPRY
Gene Name
B-box and SPRY domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317943.1 Intron NP_001304872.1
NM_001317944.1 Intron NP_001304873.1
NM_017688.2 Intron NP_060158.2
XM_006717149.3 Intron XP_006717212.1

View Full Product Details