Product Details

SNP ID
rs806429
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:88051789 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTGCCCATTATTTCACCCTGTCAT[C/T]TTTTAGATCCAGATGTATCATCAAA
Phenotype
MIM: 612739
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SPACA1 PubMed Links

Gene Details

Gene
SPACA1
Gene Name
sperm acrosome associated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030960.2 Intron NP_112222.1
XM_011536160.2 Intron XP_011534462.1
XM_017011335.1 Intron XP_016866824.1

View Full Product Details