Product Details

SNP ID
rs11234656
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.11:86443701 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
CTGTGTGTCTCAGAAGTGACAAGGC[C/T]AAGAAGTTGGTCTCCCCTCAATGTT
Phenotype
MIM: 604626
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ME3 PubMed Links

Gene Details

Gene
ME3
Gene Name
malic enzyme 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014811.1 Intron NP_001014811.1
NM_001161586.1 Intron NP_001155058.1
NM_006680.2 Intron NP_006671.2
XM_005273717.1 Intron XP_005273774.1
XM_017017136.1 Intron XP_016872625.1
XM_017017137.1 Intron XP_016872626.1
XM_017017138.1 Intron XP_016872627.1

View Full Product Details