Product Details

SNP ID
rs2054213
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.16:30960489 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTTCCCTGATCATACTCTGTACAAA[G/A]GTTTCCTTCCCTGGGCTCCAGAACA
Phenotype
MIM: 610930 MIM: 611052
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
ORAI3 PubMed Links

Gene Details

Gene
ORAI3
Gene Name
ORAI calcium release-activated calcium modulator 3
There are no transcripts associated with this gene.

Gene
SETD1A
Gene Name
SET domain containing 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014712.2 Intron NP_055527.1
XM_005255723.1 Intron XP_005255780.1
XM_006721106.3 Intron XP_006721169.1
XM_017023909.1 Intron XP_016879398.1

View Full Product Details