Product Details

SNP ID
rs13142729
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:80331448 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATGGAAAGAGGCCAGAAGAGTATA[C/G]GGAAACCAGAAAAGATTATATTCTT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C4orf22 PubMed Links

Gene Details

Gene
C4orf22
Gene Name
chromosome 4 open reading frame 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206997.1 Intron NP_001193926.1
NM_152770.2 Intron NP_689983.2
XM_011531816.2 Intron XP_011530118.2
XM_017007972.1 Intron XP_016863461.1
XM_017007973.1 Intron XP_016863462.1
XM_017007974.1 Intron XP_016863463.1
XM_017007975.1 Intron XP_016863464.1
XM_017007976.1 Intron XP_016863465.1
XM_017007977.1 Intron XP_016863466.1
XM_017007978.1 Intron XP_016863467.1

View Full Product Details