Product Details

SNP ID
rs290175
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:85695848 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTTAATACAGTGACTGGTGCTCCA[A/T]AAATGTTATTGTTATTACTCAAAGC
Phenotype
MIM: 612880
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CCDC89 PubMed Links

Gene Details

Gene
CCDC89
Gene Name
coiled-coil domain containing 89
There are no transcripts associated with this gene.

Gene
SYTL2
Gene Name
synaptotagmin like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001162951.2 Intron NP_001156423.1
NM_001162952.2 Intron NP_001156424.1
NM_001162953.2 Intron NP_001156425.1
NM_001289608.1 Intron NP_001276537.1
NM_001289609.1 Intron NP_001276538.1
NM_001289610.1 Intron NP_001276539.1
NM_032943.4 Intron NP_116561.1
NM_206929.3 Intron NP_996812.1
NM_206930.3 Intron NP_996813.1
XM_005274057.1 Intron XP_005274114.1
XM_005274060.1 Intron XP_005274117.1
XM_005274061.1 Intron XP_005274118.1
XM_005274062.2 Intron XP_005274119.1
XM_005274063.1 Intron XP_005274120.1
XM_005274066.1 Intron XP_005274123.1
XM_005274067.4 Intron XP_005274124.1
XM_011545105.1 Intron XP_011543407.1
XM_011545106.1 Intron XP_011543408.1
XM_011545107.2 Intron XP_011543409.1
XM_011545108.1 Intron XP_011543410.1
XM_011545109.2 Intron XP_011543411.1
XM_011545110.1 Intron XP_011543412.1
XM_011545112.1 Intron XP_011543414.1
XM_011545114.2 Intron XP_011543416.1
XM_011545115.1 Intron XP_011543417.1
XM_011545116.2 Intron XP_011543418.1
XM_017017933.1 Intron XP_016873422.1
XM_017017934.1 Intron XP_016873423.1
XM_017017935.1 Intron XP_016873424.1
XM_017017936.1 Intron XP_016873425.1
XM_017017937.1 Intron XP_016873426.1
XM_017017938.1 Intron XP_016873427.1
XM_017017939.1 Intron XP_016873428.1
XM_017017940.1 Intron XP_016873429.1

View Full Product Details