Product Details

SNP ID
rs1969269
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:47258171 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGTTTTACCCAGCCCTCTCTGCTG[A/C]CCACAGAGTGGGCTGCTCCACCCCA
Phenotype
MIM: 173470
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ITGB3 PubMed Links

Gene Details

Gene
ITGB3
Gene Name
integrin subunit beta 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000212.2 Intron NP_000203.2

View Full Product Details