Product Details

SNP ID
rs34000404
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:20696465 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGTACATCTGCGGGATCTAACTA[C/T]AACCTTCCTAAGCCACTTGCTCTCT
Phenotype
MIM: 613389
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLCO1C1 PubMed Links

Gene Details

Gene
SLCO1C1
Gene Name
solute carrier organic anion transporter family member 1C1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145944.1 Intron NP_001139416.1
NM_001145945.1 Intron NP_001139417.1
NM_001145946.1 Intron NP_001139418.1
NM_017435.4 Intron NP_059131.1
XM_005253394.2 Intron XP_005253451.1
XM_005253396.2 Intron XP_005253453.1
XM_011520703.2 Intron XP_011519005.1
XM_011520704.2 Intron XP_011519006.1
XM_011520709.2 Intron XP_011519011.1
XM_011520711.2 Intron XP_011519013.1
XM_017019484.1 Intron XP_016874973.1
XM_017019485.1 Intron XP_016874974.1
XM_017019486.1 Intron XP_016874975.1
XM_017019487.1 Intron XP_016874976.1
XM_017019488.1 Intron XP_016874977.1
XM_017019489.1 Intron XP_016874978.1
XM_017019490.1 Intron XP_016874979.1

View Full Product Details