Product Details
- SNP ID
-
rs9919
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:123621292 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AATGTGCTTACCACTGGAAGCCAGA[C/T]GCAGATTCAGCAAGTGTTTCTTGCC
- Phenotype
-
MIM: 606686
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DDX55
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs113041500] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DDX55
- Gene Name
- DEAD-box helicase 55
- Gene
- EIF2B1
- Gene Name
- eukaryotic translation initiation factor 2B subunit alpha
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001414.3 |
1601 |
UTR 3 |
|
|
NP_001405.1 |
View Full Product Details