Product Details

SNP ID
rs2184334
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.10:70211798 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CATGCCAAGTTCTCTTGCATAAGAA[A/G]CACGTTGGAAATGCACACACGGACC
Phenotype
MIM: 179030
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PPA1 PubMed Links

Gene Details

Gene
PPA1
Gene Name
pyrophosphatase (inorganic) 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021129.3 Intron NP_066952.1

View Full Product Details