Product Details

SNP ID
rs3746455
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.20:34369410 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTTTCTCAGGTACCATGCATTTCAC[A/G]GTGGCCTTGTGGAGACAACGCCTTA
Phenotype
MIM: 606409
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ITCH PubMed Links

Gene Details

Gene
ITCH
Gene Name
itchy E3 ubiquitin protein ligase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001257137.2 132 UTR 5 NP_001244066.1
NM_001257138.2 132 UTR 5 NP_001244067.1
NM_001324197.1 132 UTR 5 NP_001311126.1
NM_001324198.1 132 UTR 5 NP_001311127.1
NM_031483.6 132 UTR 5 NP_113671.3
XM_005260570.1 132 UTR 5 XP_005260627.1
XM_011529079.1 132 Intron XP_011527381.1
XM_017028089.1 132 Intron XP_016883578.1
XM_017028090.1 132 UTR 5 XP_016883579.1
XM_017028091.1 132 UTR 5 XP_016883580.1
XM_017028092.1 132 UTR 5 XP_016883581.1
Gene
LOC107985381
Gene Name
uncharacterized LOC107985381
There are no transcripts associated with this gene.

View Full Product Details