Product Details

SNP ID
rs966365
Assay Type
Validated
NCBI dbSNP Submissions
77
Location
Chr.1:236394471 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TGGGCCTCAGGACGACTAAACAGAT[A/G]GGGAGAGGCACTAAAGCTCCTGGTC
Phenotype
MIM: 606603
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EDARADD PubMed Links

Gene Details

Gene
EDARADD
Gene Name
EDAR associated death domain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080738.3 92 Intron NP_542776.1
NM_145861.2 92 Missense Mutation ATA,ATG I9M NP_665860.2

View Full Product Details