Product Details

SNP ID
rs7513444
Assay Type
Functionally Tested
NCBI dbSNP Submissions
39
Location
Chr.1:206573713 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCCGAGATGGATGGTCTACTTGGA[A/G]GTTAATATAAGTAGCCAGGCGTAGT
Phenotype
MIM: 613709 MIM: 607020
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
EIF2D PubMed Links

Gene Details

Gene
EIF2D
Gene Name
eukaryotic translation initiation factor 2D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201478.1 Intron NP_001188407.1
NM_006893.2 Intron NP_008824.2
XM_005277509.3 Intron XP_005277566.1
XM_006711196.3 Intron XP_006711259.1
XM_011509257.2 Intron XP_011507559.1
XM_011509258.2 Intron XP_011507560.1
XM_017000530.1 Intron XP_016856019.1
XM_017000531.1 Intron XP_016856020.1
XM_017000532.1 Intron XP_016856021.1
Gene
RASSF5
Gene Name
Ras association domain family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182663.3 Intron NP_872604.1
NM_182664.3 Intron NP_872605.1
NM_182665.3 Intron NP_872606.1

View Full Product Details