Product Details

SNP ID
rs1413100
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:165280135 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTTAAAATACATAAGAAATGAATA[C/T]CATAGGTTAGAAAAAAATACATTAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C6orf118 PubMed Links

Gene Details

Gene
C6orf118
Gene Name
chromosome 6 open reading frame 118
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144980.3 Intron NP_659417.2
XM_005266838.2 Intron XP_005266895.1
XM_011535509.2 Intron XP_011533811.1
XM_011535510.2 Intron XP_011533812.1
XM_011535511.2 Intron XP_011533813.1
XM_011535512.2 Intron XP_011533814.1
XM_011535513.2 Intron XP_011533815.1
XM_017010323.1 Intron XP_016865812.1
XM_017010324.1 Intron XP_016865813.1

View Full Product Details