Product Details

SNP ID
rs2811907
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:83629444 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGGATCAAGACCAAATACTTACAC[C/T]GCTAAGCATGCATGGCATGGCCTCC
Phenotype
MIM: 611343
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IDNK PubMed Links

Gene Details

Gene
IDNK
Gene Name
IDNK, gluconokinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001551.3 Intron NP_001001551.2
NM_001256915.1 Intron NP_001243844.1
XM_006717110.3 Intron XP_006717173.1
XM_006717111.3 Intron XP_006717174.1
XM_017014719.1 Intron XP_016870208.1
XM_017014720.1 Intron XP_016870209.1
XM_017014721.1 Intron XP_016870210.1
XM_017014722.1 Intron XP_016870211.1
XM_017014723.1 Intron XP_016870212.1
XM_017014724.1 Intron XP_016870213.1
XM_017014725.1 Intron XP_016870214.1

View Full Product Details