Product Details

SNP ID
rs843540
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:196748442 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAACATGTCTCCACCAGTATCATAC[A/C]GACTGGTTTCCGCACCTTAAAACTC
Phenotype
MIM: 605022
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PAK2 PubMed Links

Gene Details

Gene
PAK2
Gene Name
p21 (RAC1) activated kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002577.4 Intron NP_002568.2
XM_011512870.2 Intron XP_011511172.1
XM_017006501.1 Intron XP_016861990.1

View Full Product Details